accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 53213-83-5

IUPAC Name: 3-heptyl-2-[3-(3-heptyl-2,3-dihydro-1,3-benzoxazol-2-ylidene)prop-1-en-1-yl]-1,3-benzoxazol-3-ium iodide
Synonyms: 3-heptyl-2-[3-(3-heptyl-1,3-benzoxazol-2-ylidene)prop-1-en-1-yl]-1,3-benzoxazol-3-ium iodide
Molecular Weight (g/mol): 600.59
Molecular Formula: C31H41IN2O2
InChi Key: ANJZLSIIEGUCQL-UHFFFAOYSA-M
SMILES: [I-].CCCCCCCN1C(OC2=CC=CC=C12)=CC=CC1=[N+](CCCCCCC)C2=CC=CC=C2O1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results