accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 53222-92-7

IUPAC Name: 3-amino-2-methylphenol
Synonyms: 3-amino-2-methylphenol
Molecular Weight (g/mol): 123.16
Molecular Formula: C7H9NO
InChi Key: FLROJJGKUKLCAE-UHFFFAOYSA-N
SMILES: CC1=C(N)C=CC=C1O
Molecular Weight (g/mol) 
  • (2)
  • (3)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results