accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NaH3CSOOO

CAS RN 5324-84-5

IUPAC Name: sodium octane-1-sulfonate
Synonyms: sodium 1-octanesulfonate sodium caprylyl sulfonate
Molecular Weight (g/mol): 216.27
Molecular Formula: C8H17NaO3S
InChi Key: HRQDCDQDOPSGBR-UHFFFAOYSA-M
SMILES: [Na+].CCCCCCCCS([O-])(=O)=O
Quantity 
  • (7)
  • (8)
  • (4)
  • (1)
  • (1)
Physical Form 
  • (3)
  • (7)
  • (1)
  • (3)
Percent Purity 
  • (5)
  • (3)
  • (2)
Grade 
  • (1)
  • (4)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (4)
  • (6)
  • (1)
  • (6)
  • (3)
  • (3)
  • (5)
  • (7)
  • (8)
  • (4)
  • (1)
  • (1)
  • (3)
  • (7)
  • (1)
  • (3)
  • (5)
  • (3)
  • (2)
  • (1)
  • (4)
  • (2)