accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 53250-83-2

CH3SOOOOCl

CAS RN 53250-83-2

IUPAC Name: 2-chloro-4-methanesulfonylbenzoate
Synonyms: 2-chloro-4-methanesulfonylbenzoate
Molecular Weight (g/mol): 233.64
Molecular Formula: C8H6ClO4S
InChi Key: CTTWSFIIFMWHLQ-UHFFFAOYSA-M
SMILES: CS(=O)(=O)C1=CC=C(C([O-])=O)C(Cl)=C1
Percent Purity 
  • (5)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (3)
  • (5)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)