accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3ONHH3CN

CAS RN 5327-33-3

IUPAC Name: N-(6-methylpyridin-2-yl)acetamide
Synonyms: N-(6-methylpyridin-2-yl)acetamide
Molecular Weight (g/mol): 150.18
Molecular Formula: C8H10N2O
InChi Key: UEXYAZLLFZIXHN-UHFFFAOYSA-N
SMILES: CC(=O)NC1=CC=CC(C)=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results