accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 533-31-3

IUPAC Name: 2H-1,3-benzodioxol-5-ol
Synonyms: sesamol sesamol, ion (1+) sesamol sodium sesamol titanium (+4) ion (1+) sesamol methylene ether of oxyhydroquinone sesamol lithium
Molecular Weight (g/mol): 138.12
Molecular Formula: C7H6O3
InChi Key: LUSZGTFNYDARNI-UHFFFAOYSA-N
SMILES: OC1=CC=C2OCOC2=C1
Molecular Weight (g/mol) 
  • (8)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
Boiling Point 
  • (3)
Certifications/Compliance 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results