accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NH2NF

CAS RN 53312-81-5

IUPAC Name: 5-amino-2-fluorobenzonitrile
Synonyms: 5-amino-2-fluorobenzonitrile
Molecular Weight (g/mol): 136.13
Molecular Formula: C7H5FN2
InChi Key: HHTRAISBAAXRKZ-UHFFFAOYSA-N
SMILES: NC1=CC(C#N)=C(F)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results