accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 5333-86-8

IUPAC Name: hydrogen ethyl benzenecarboximidate chloride
Synonyms: hydrogen ethyl benzenecarboximidate chloride
Molecular Weight (g/mol): 185.65
Molecular Formula: C9H12ClNO
InChi Key: MODZVIMSNXSQIH-UHFFFAOYSA-N
SMILES: [H+].[Cl-].CCOC(=N)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results