accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OHONHNNOO

CAS RN 5334-40-7

IUPAC Name: 4-nitro-1H-pyrazole-5-carboxylic acid
Synonyms: 4-nitro-2H-pyrazole-3-carboxylic acid
Molecular Weight (g/mol): 157.09
Molecular Formula: C4H3N3O4
InChi Key: ZMAXXOYJWZZQBK-UHFFFAOYSA-N
SMILES: OC(=O)C1=C(C=NN1)[N+]([O-])=O
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results