accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 53344-74-4

IUPAC Name: 2,2'-dichloro-4,4'-bipyridine
Synonyms: 2,2'-dichloro-4,4'-bipyridine
Molecular Weight (g/mol): 225.07
Molecular Formula: C10H6Cl2N2
InChi Key: KSELKNWXBOXHEV-UHFFFAOYSA-N
SMILES: ClC1=NC=CC(=C1)C1=CC(Cl)=NC=C1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results