CAS RN 5339-69-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
N-Isopropylbenzenesulfonamide, TRC
CAS: 5339-69-5 Molecular Formula: C9H13NO2S Molecular Weight (g/mol): 199.27 Synonym: NSC 3549,N-(1-Methylethyl)benzenesulfonamide,Isopropylbenzenesulfonamide,N-Benzenesulfonyl-N-isopropylamine IUPAC Name: N-propan-2-ylbenzenesulfonamide SMILES: CC(C)NS(=O)(=O)c1ccccc1