accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3NHSNHH3C

CAS RN 534-13-4

IUPAC Name: 1,3-dimethylthiourea
Synonyms: N,N-dimethylthiourea
Molecular Weight (g/mol): 104.17
Molecular Formula: C3H8N2S
InChi Key: VLCDUOXHFNUCKK-UHFFFAOYSA-N
SMILES: CNC(=S)NC
Molecular Weight (g/mol) 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)