accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 53408-94-9

IUPAC Name: λ²-tin(2+) dimethanesulfonate
Synonyms: λ²-tin(2+) dimesylate
Molecular Weight (g/mol): 308.89
Molecular Formula: C2H6O6S2Sn
InChi Key: JALQQBGHJJURDQ-UHFFFAOYSA-L
SMILES: [Sn++].CS([O-])(=O)=O.CS([O-])(=O)=O
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results