accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OPOOOOPOOOMgMgMg

CAS RN 53408-95-0

IUPAC Name: trimagnesium(2+) hydrate diphosphate
Synonyms: trimagnesium(2+) hydrate diphosphate
Molecular Weight (g/mol): 280.87
Molecular Formula: H2Mg3O9P2
InChi Key: WESIJKDWGUWFEP-UHFFFAOYSA-H
SMILES: O.[Mg++].[Mg++].[Mg++].[O-]P([O-])([O-])=O.[O-]P([O-])([O-])=O
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results