accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H2NNHOHO

CAS RN 5341-58-2

IUPAC Name: 3-hydroxynaphthalene-2-carbohydrazide
Synonyms: 3-hydroxynaphthalene-2-carbohydrazide
Molecular Weight (g/mol): 202.21
Molecular Formula: C11H10N2O2
InChi Key: FDNAQCWUERCJBK-UHFFFAOYSA-N
SMILES: NNC(=O)C1=C(O)C=C2C=CC=CC2=C1
Molecular Weight (g/mol) 
  • (3)
  • (3)
Percent Purity 
  • (1)
  • (5)
Quantity 
  • (2)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (2)
  • (3)
  • (3)
  • (3)
  • (1)
  • (5)
  • (2)
  • (2)
  • (2)
  • (1)