accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

HClNH2H2N

CAS RN 5341-61-7

IUPAC Name: hydrogen hydrazine chloride
Synonyms: hydrazine dihydrochloride
Molecular Weight (g/mol): 68.5
Molecular Formula: ClH5N2
InChi Key: BIVUUOPIAYRCAP-UHFFFAOYSA-N
SMILES: [H+].[Cl-].NN
Molecular Weight (g/mol) 
  • (3)
  • (6)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
Percent Purity 
  • (3)
  • (3)
Physical Form 
  • (3)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (2)
  • (3)
  • (3)
  • (6)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (3)
  • (3)
  • (2)
  • (1)