accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COBrNOO

CAS RN 5344-78-5

IUPAC Name: 1-bromo-4-methoxy-2-nitrobenzene
Synonyms: 1-bromo-4-methoxy-2-nitrobenzene
Molecular Weight (g/mol): 232.03
Molecular Formula: C7H6BrNO3
InChi Key: KCOBIBRGPCFIGF-UHFFFAOYSA-N
SMILES: COC1=CC=C(Br)C(=C1)[N+]([O-])=O
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results