accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 5346-38-3

IUPAC Name: pyridine-2-carbothioamide
Synonyms: 2-pyridinethiocarboxamide
Molecular Weight (g/mol): 138.19
Molecular Formula: C6H6N2S
InChi Key: HYKQYVSNFPWGKQ-UHFFFAOYSA-N
SMILES: NC(=S)C1=CC=CC=N1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results