accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

HClCH3NOHN

CAS RN 5348-51-6

IUPAC Name: hydrogen 6-methyl-1,2-dihydropyrimidin-2-one chloride
Synonyms: hydrogen 2-pyrimidone,6-methyl chloride
Molecular Weight (g/mol): 146.57
Molecular Formula: C5H7ClN2O
InChi Key: FDRYQDWXFZBBAD-UHFFFAOYSA-N
SMILES: [H+].[Cl-].CC1=CC=NC(=O)N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results