accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COOCH3Br

CAS RN 535-11-5

IUPAC Name: ethyl 2-bromopropanoate
Synonyms: ethyl a-bromopropionate
Molecular Weight (g/mol): 181.03
Molecular Formula: C5H9BrO2
InChi Key: ARFLASKVLJTEJD-UHFFFAOYNA-N
SMILES: CCOC(=O)C(C)Br
Quantity 
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results