accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 53531-31-0

FFFO

CAS RN 53531-31-0

IUPAC Name: 1-(anthracen-9-yl)-2,2,2-trifluoroethan-1-one
Synonyms: 1-(anthracen-9-yl)-2,2,2-trifluoroethanone
Molecular Weight (g/mol): 274.24
Molecular Formula: C16H9F3O
InChi Key: MNCMBBIFTVWHIP-UHFFFAOYSA-N
SMILES: FC(F)(F)C(=O)C1=C2C=CC=CC2=CC2=CC=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results