accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 53588-95-7

H3CH3CCH3OONHN

CAS RN 53588-95-7

IUPAC Name: tert-butyl N-(2-cyanoethyl)carbamate
Synonyms: tert-butyl N-(2-cyanoethyl)carbamate
Molecular Weight (g/mol): 170.21
Molecular Formula: C8H14N2O2
InChi Key: NORLFIHQJFOIGS-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)NCCC#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)