accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3NCH3SSHNO(Z)

CAS RN 536-17-4

IUPAC Name: (5Z)-5-{[4-(dimethylamino)phenyl]methylidene}-2-sulfanylidene-1,3-thiazolidin-4-one
Synonyms: (5Z)-5-{[4-(dimethylamino)phenyl]methylidene}-2-sulfanylidene-1,3-thiazolidin-4-one
Molecular Weight (g/mol): 264.36
Molecular Formula: C12H12N2OS2
InChi Key: JJRVRELEASDUMY-YFHOEESVSA-N
SMILES: CN(C)C1=CC=C(\C=C2/SC(=S)NC2=O)C=C1
Molecular Weight (g/mol) 
  • (3)
  • (1)
  • (4)
  • (3)
Quantity 
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
Percent Purity 
  • (3)
  • (3)
Physical Form 
  • (3)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (3)
  • (3)
  • (1)
  • (3)
  • (1)
  • (4)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)