accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NH2HNOCl

CAS RN 536-40-3

IUPAC Name: 4-chlorobenzohydrazide
Synonyms: benzoylhydrazine, P-chloro
Molecular Weight (g/mol): 170.6
Molecular Formula: C7H7ClN2O
InChi Key: PKBGHORNUFQAAW-UHFFFAOYSA-N
SMILES: NNC(=O)C1=CC=C(Cl)C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results