accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CCH3OHO

CAS RN 536-66-3

IUPAC Name: 4-(propan-2-yl)benzoic acid
Synonyms: 4-isopropylbenzoic acid cumic acid cuminic acid
Molecular Weight (g/mol): 164.2
Molecular Formula: C10H12O2
InChi Key: CKMXAIVXVKGGFM-UHFFFAOYSA-N
SMILES: CC(C)C1=CC=C(C=C1)C(O)=O
Molecular Weight (g/mol) 
  • (5)
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)
  • (1)
Percent Purity 
  • (4)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (3)
  • (4)
  • (5)
  • (4)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)
  • (1)
  • (4)
  • (2)
  • (2)