accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 536-74-3

IUPAC Name: ethynylbenzene
Synonyms: phenylacetylene
Molecular Weight (g/mol): 102.14
Molecular Formula: C8H6
InChi Key: UEXCJVNBTNXOEH-UHFFFAOYSA-N
SMILES: C#CC1=CC=CC=C1
Molecular Weight (g/mol) 
  • (3)
  • (3)
  • (3)
Quantity 
  • (1)
  • (3)
  • (2)
  • (1)
  • (2)
Boiling Point 
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (3)
Grade 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (3)
  • (2)
  • (1)
  • (2)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)