accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 536-89-0

CH3HNNH2

CAS RN 536-89-0

IUPAC Name: (3-methylphenyl)hydrazine
Synonyms: (3-methylphenyl)hydrazine
Molecular Weight (g/mol): 122.17
Molecular Formula: C7H10N2
InChi Key: GPTOGZLZMLJZCV-UHFFFAOYSA-N
SMILES: CC1=CC=CC(NN)=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results