accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 5368-81-0

CH3OOOCH3

CAS RN 5368-81-0

IUPAC Name: methyl 3-methoxybenzoate
Synonyms: methyl 3-methoxybenzoate
Molecular Weight (g/mol): 166.18
Molecular Formula: C9H10O3
InChi Key: DUKYPQBGYRJVAN-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC=CC(OC)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results