accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 5373-87-5

IUPAC Name: (E)-N'-hydroxy-4-methoxybenzene-1-carboximidamide
Synonyms: (E)-N'-hydroxy-4-methoxybenzenecarboximidamide
Molecular Weight (g/mol): 166.18
Molecular Formula: C8H10N2O2
InChi Key: WVALRFKCJCIVBR-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=C1)C(\N)=N/O
Quantity 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results