accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NH2NNNH2(E/Z)

CAS RN 538-41-0

IUPAC Name: 4-[2-(4-aminophenyl)diazen-1-yl]aniline
Synonyms: 4-[2-(4-aminophenyl)diazen-1-yl]aniline
Molecular Weight (g/mol): 212.26
Molecular Formula: C12H12N4
InChi Key: KQIKKETXZQDHGE-UHFFFAOYSA-N
SMILES: NC1=CC=C(C=C1)N=NC1=CC=C(N)C=C1
Molecular Weight (g/mol) 
  • (3)
  • (5)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (5)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)