accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3C

CAS RN 538-68-1

IUPAC Name: pentylbenzene
Synonyms: pentylbenzene
Molecular Weight (g/mol): 148.25
Molecular Formula: C11H16
InChi Key: PWATWSYOIIXYMA-UHFFFAOYSA-N
SMILES: CCCCCC1=CC=CC=C1
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)