accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 538-74-9

IUPAC Name: [(benzylsulfanyl)methyl]benzene
Synonyms: benzyl sulfide
Molecular Weight (g/mol): 214.33
Molecular Formula: C14H14S
InChi Key: LUFPJJNWMYZRQE-UHFFFAOYSA-N
SMILES: C(SCC1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results