accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NC(?)N

CAS RN 538-75-0

IUPAC Name: N,N'-dicyclohexylmethanediimine
Synonyms: dicyclohexylcarbodiimide DCC carbodicyclohexylimide DCCI DCCD
Molecular Weight (g/mol): 206.33
Molecular Formula: C13H22N2
InChi Key: QOSSAOTZNIDXMA-UHFFFAOYSA-N
SMILES: C1CCC(CC1)N=C=NC1CCCCC1
Molecular Weight (g/mol) 
  • (9)
  • (3)
Quantity 
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
Percent Purity 
  • (3)
  • (8)
Boiling Point 
  • (5)
  • (3)
Physical Form 
  • (5)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (9)
  • (3)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (8)
  • (5)
  • (3)
  • (5)
  • (1)