accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3O

CAS RN 538-86-3

IUPAC Name: (methoxymethyl)benzene
Synonyms: benzyl methyl ether
Molecular Weight (g/mol): 122.17
Molecular Formula: C8H10O
InChi Key: GQKZBCPTCWJTAS-UHFFFAOYSA-N
SMILES: COCC1=CC=CC=C1
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results