accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 53801-63-1

IUPAC Name: copper zinc
Synonyms: copper zinc
Molecular Weight (g/mol): 128.93
Molecular Formula: CuZn
InChi Key: TVZPLCNGKSPOJA-UHFFFAOYSA-N
SMILES: [Cu].[Zn]
Quantity 
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results