accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 53817-44-0

CH3H3CH3CNOHCH3

CAS RN 53817-44-0

IUPAC Name: 2-(2,2,4,7-tetramethyl-1,2,3,4-tetrahydroquinolin-1-yl)ethan-1-ol
Synonyms: 2-(2,2,4,7-tetramethyl-3,4-dihydroquinolin-1-yl)ethanol
Molecular Weight (g/mol): 233.36
Molecular Formula: C15H23NO
InChi Key: DICAWUKDDRKZDZ-UHFFFAOYNA-N
SMILES: CC1CC(C)(C)N(CCO)C2=CC(C)=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results