accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 5382-23-0

HClCH3NCl

CAS RN 5382-23-0

IUPAC Name: hydrogen 4-chloro-1-methylpiperidine chloride
Synonyms: hydrogen 4-chloro-1-methylpiperidine chloride
Molecular Weight (g/mol): 170.08
Molecular Formula: C6H13Cl2N
InChi Key: QDOJNWFOCOWAPO-UHFFFAOYSA-N
SMILES: [H+].[Cl-].CN1CCC(Cl)CC1
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)