accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OONOO

CAS RN 5383-82-4

IUPAC Name: 4-(1,3-dioxo-2,3-dihydro-1H-isoindol-2-yl)benzoate
Synonyms: 4-(1,3-dioxoisoindol-2-yl)benzoate
Molecular Weight (g/mol): 266.23
Molecular Formula: C15H8NO4
InChi Key: HBXCMGPWVGKNRS-UHFFFAOYSA-M
SMILES: [O-]C(=O)C1=CC=C(C=C1)N1C(=O)C2=CC=CC=C2C1=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results