accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COCH3H3CN

CAS RN 53833-30-0

IUPAC Name: 2-ethyl-4,5-dimethyl-1,3-oxazole
Synonyms: 2-ethyl-4,5-dimethyl-1,3-oxazole
Molecular Weight (g/mol): 125.17
Molecular Formula: C7H11NO
InChi Key: LCYOFVYHDBWYSI-UHFFFAOYSA-N
SMILES: CCC1=NC(C)=C(C)O1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results