accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 53846-29-0

IUPAC Name: tris(λ²-lead(2+)) bis(1,3,5-triazine-2,4,6-tris(olate)) trihydrate
Synonyms: tris(λ²-lead(2+)) bis(1,3,5-triazine-2,4,6-tris(olate)) trihydrate
Molecular Weight (g/mol): 927.7
Molecular Formula: C6H6N6O9Pb3
InChi Key: DPLKIANDZYQWKF-UHFFFAOYSA-H
SMILES: O.O.O.[Pb++].[Pb++].[Pb++].[O-]C1=NC([O-])=NC([O-])=N1.[O-]C1=NC([O-])=NC([O-])=N1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results