accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

BrHNN

CAS RN 53857-57-1

IUPAC Name: 5-bromo-1H-indazole
Synonyms: 5-bromo-1H-indazole
Molecular Weight (g/mol): 197.04
Molecular Formula: C7H5BrN2
InChi Key: STVHMYNPQCLUNJ-UHFFFAOYSA-N
SMILES: BrC1=CC=C2NN=CC2=C1
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results