accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 539-47-9

HOOO(E)

CAS RN 539-47-9

IUPAC Name: (2E)-3-(furan-2-yl)prop-2-enoic acid
Synonyms: (2E)-3-(furan-2-yl)prop-2-enoic acid
Molecular Weight (g/mol): 138.12
Molecular Formula: C7H6O3
InChi Key: ZCJLOOJRNPHKAV-ONEGZZNKSA-N
SMILES: OC(=O)\C=C\C1=CC=CO1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results