accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H2NNH2

CAS RN 539-48-0

IUPAC Name: 1-[4-(aminomethyl)phenyl]methanamine
Synonyms: 1-[4-(aminomethyl)phenyl]methanamine
Molecular Weight (g/mol): 136.2
Molecular Formula: C8H12N2
InChi Key: ISKQADXMHQSTHK-UHFFFAOYSA-N
SMILES: NCC1=CC=C(CN)C=C1
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results