accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 539-80-0

O

CAS RN 539-80-0

IUPAC Name: cyclohepta-2,4,6-trien-1-one
Synonyms: tropone
Molecular Weight (g/mol): 106.12
Molecular Formula: C7H6O
InChi Key: QVWDCTQRORVHHT-UHFFFAOYSA-N
SMILES: O=C1C=CC=CC=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)