accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COOCH3O

CAS RN 539-88-8

IUPAC Name: ethyl 4-oxopentanoate
Synonyms: ethyl levulinate
Molecular Weight (g/mol): 144.17
Molecular Formula: C7H12O3
InChi Key: GMEONFUTDYJSNV-UHFFFAOYSA-N
SMILES: CCOC(=O)CCC(C)=O
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)
Boiling Point 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)
  • (3)
  • (2)
  • (2)