accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 539-93-5

H3COOOHOOCH3

CAS RN 539-93-5

IUPAC Name: 3-(dodecanoyloxy)-2-hydroxypropyl dodecanoate
Synonyms: 3-(dodecanoyloxy)-2-hydroxypropyl dodecanoate
Molecular Weight (g/mol): 456.71
Molecular Formula: C27H52O5
InChi Key: KUVAEMGNHJQSMH-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCC(=O)OCC(O)COC(=O)CCCCCCCCCCC
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results