accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NH2NNH2NH2H2NNOHSOHOO

CAS RN 5392-28-9

IUPAC Name: pyrimidine-2,4,5,6-tetramine; sulfuric acid
Synonyms: tetraaminopyrimidine sulfate
Molecular Weight (g/mol): 238.22
Molecular Formula: C4H10N6O4S
InChi Key: MQEFDQWUCTUJCP-UHFFFAOYSA-N
SMILES: OS(O)(=O)=O.NC1=NC(N)=C(N)C(N)=N1
Molecular Weight (g/mol) 
  • (7)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
Percent Purity 
  • (3)
  • (2)
Certifications/Compliance 
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (3)
  • (7)
  • (3)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)