accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3CH3CH3O(E)

CAS RN 5392-40-5

IUPAC Name: (2E)-3,7-dimethylocta-2,6-dienal
Synonyms: α-citral citral A citral B citral α epa pesticide chemical code 040510 fema number 2303 citral (natural) β-geranial geranial 2,6-dimethyloctadien-2,6-al-8
Molecular Weight (g/mol): 152.24
Molecular Formula: C10H16O
InChi Key: WTEVQBCEXWBHNA-JXMROGBWSA-N
SMILES: CC(C)=CCC\C(C)=C\C=O
Molecular Weight (g/mol) 
  • (4)
  • (6)
Percent Purity 
  • (3)
  • (6)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (3)
  • (3)
  • (4)
  • (4)
  • (6)
  • (3)
  • (6)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)