accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CO(Z)

CAS RN 53939-28-9

IUPAC Name: (11Z)-hexadec-11-enal
Synonyms: (11Z)-hexadec-11-enal
Molecular Weight (g/mol): 238.42
Molecular Formula: C16H30O
InChi Key: AMTITFMUKRZZEE-WAYWQWQTSA-N
SMILES: CCCC\C=C/CCCCCCCCCC=O
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results