CAS RN 53949-54-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
Hydroxy Ibuprofen, TRC
CAS: 53949-54-5 Molecular Formula: C13 H18 O3 Molecular Weight (g/mol): 222.28 Synonym: Benzeneacetic acid, 4-(3-hydroxy-2-methylpropyl)-α-methyl-,2-[4-(3-Hydroxy-2-methyl-propyl)phenyl]propanoic acid,4-(3-Hydroxy-2-methylpropyl)-α-methylbenzeneacetic acid,2,4'-(2-Hydroxymethylpropyl)phenylpropionic acid,2-[4-(2-Hydroxymethylpropyl)phenyl]propanoic acid,3-Hydroxyibuprofen IUPAC Name: 2-[4-(3-hydroxy-2-methylpropyl)phenyl]propanoic acid SMILES: CC(CO)Cc1ccc(cc1)C(C)C(=O)O